3-(tert-Butyl)-4-isopropoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

63682-500MG 63682-1G 63682-5G 63682-25G
77331-964EA 184.37 USD
77331-964 77347-260 77368-064 77389-404
3-(tert-Butyl)-4-isopropoxybenzaldehyde ≥97%
3-(tert-Butyl)-4-isopropoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₁₄H₂₀O₂
MW: 220.3074 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29122392
CAS Number: 1289169-85-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR