3-(tert-Butyl)-4-isopropoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

63679-500MG 63679-1G 63679-5G 63679-25G
77331-962EA 260.59 USD
77331-962 77356-310 77368-062 77389-402
3-(tert-Butyl)-4-isopropoxybenzoic acid ≥97%
3-(tert-Butyl)-4-isopropoxybenzoic acid

Precaución: for Research Use Only

Formula: C₁₄H₂₀O₃
Storage Temperature: Ambient
MDL Number: MFCD19309097
CAS Number: 142651-36-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR