(3-(tert-Butyl)-4-ethoxyphenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

63675-500MG 63675-1G 63675-5G
77339-172EA 557.23 USD
77339-172 77356-304 77380-312
(3-(tert-Butyl)-4-ethoxyphenyl)(methyl)sulfane ≥97%
(3-(tert-Butyl)-4-ethoxyphenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₁₃H₂₀OS
Storage Temperature: Ambient
MDL Number: MFCD34761477
CAS Number: 2807445-10-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR