4-Bromo-2-(tert-butyl)-1-isopropoxybenzene ≥97%

Supplier: AOBChem USA

Warning

63661-1G 63661-5G 63661-25G
77341-832EA 105.06 USD
77341-832 77368-058 77380-306
4-Bromo-2-(tert-butyl)-1-isopropoxybenzene ≥97%
4-Bromo-2-(tert-butyl)-1-isopropoxybenzene

Precaución: for Research Use Only

Formula: C₁₃H₁₉BrO
MW: 271.1933 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28356571
CAS Number: 1369855-94-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR