4-Bromo-2-(tert-butyl)-1-ethoxybenzene ≥97%

Supplier: AOBChem USA

Warning

63660-500MG 63660-1G 63660-5G 63660-25G
77331-954EA 178.19 USD
77331-954 77346-614 77368-056 77389-398
4-Bromo-2-(tert-butyl)-1-ethoxybenzene ≥97%
4-Bromo-2-(tert-butyl)-1-ethoxybenzene

Precaución: for Research Use Only

Formula: C₁₂H₁₇BrO
MW: 257.1667 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32219312
CAS Number: 2713449-18-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR