1-Bromo-3-ethyl-2-fluorobenzene ≥95%

Supplier: AOBChem USA

Warning

63654-500MG 63654-1G 63654-5G
77339-794EA 643.75 USD
77339-794 77356-298 77380-304
1-Bromo-3-ethyl-2-fluorobenzene ≥95%
1-Bromo-3-ethyl-2-fluorobenzene

Precaución: for Research Use Only

Formula: C₈H₈BrF
Storage Temperature: Ambient
MDL Number: MFCD22490800
CAS Number: 1369914-39-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR