2,2,2-trifluoro-1-(5-(methoxymethoxy)-2-methylphenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

63629-500MG 63629-1G 63629-5G 63629-25G
77331-952EA 182.31 USD
77331-952 77346-986 77368-054 77389-396
2,2,2-trifluoro-1-(5-(methoxymethoxy)-2-methylphenyl)ethanone ≥95%
2,2,2-trifluoro-1-(5-(methoxymethoxy)-2-methylphenyl)ethanone

Precaución: for Research Use Only

Formula: C₁₁H₁₁F₃O₃
MW: 248.1984 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34761459
CAS Number: 2807444-62-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR