1-(2-(Benzyloxy)-6-chlorophenyl)-2,2,2-trifluoroethanone ≥97%

Supplier: AOBChem USA

Warning

63618-500MG 63618-1G 63618-5G
77338-084EA 462.47 USD
77338-084 77356-296 77380-302
1-(2-(Benzyloxy)-6-chlorophenyl)-2,2,2-trifluoroethanone ≥97%
1-(2-(Benzyloxy)-6-chlorophenyl)-2,2,2-trifluoroethanone

Precaución: for Research Use Only

Formula: C₁₅H₁₀ClF₃O₂
MW: 314.6869 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34761455
CAS Number: 2918856-05-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR