1-(2-Chloro-6-hydroxyphenyl)-2,2,2-trifluoroethanone ≥97%

Supplier: AOBChem USA

Warning

63615-500MG 63615-1G 63615-5G
77338-334EA 477.92 USD
77338-334 77356-294 77380-300
1-(2-Chloro-6-hydroxyphenyl)-2,2,2-trifluoroethanone ≥97%
1-(2-Chloro-6-hydroxyphenyl)-2,2,2-trifluoroethanone

Precaución: for Research Use Only

Formula: C₈H₄ClF₃O₂
MW: 224.5643 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34761452
CAS Number: 2229402-28-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR