1-(Benzyloxy)-3-chloro-2-iodobenzene ≥97%

Supplier: AOBChem USA

Warning

63604-500MG 63604-1G 63604-5G 63604-25G
77331-946EA 179.22 USD
77331-946 77346-736 77368-048 77389-390
1-(Benzyloxy)-3-chloro-2-iodobenzene ≥97%
1-(Benzyloxy)-3-chloro-2-iodobenzene

Precaución: for Research Use Only

Formula: C₁₃H₁₀ClIO
MW: 344.5753 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30295978
CAS Number: 945470-01-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR