1-Chloro-3-ethoxy-2-iodobenzene ≥97%

Supplier: AOBChem USA

Warning

63603-500MG 63603-1G 63603-5G 63603-25G
77331-944EA 179.22 USD
77331-944 77346-734 77368-046 77389-388
1-Chloro-3-ethoxy-2-iodobenzene ≥97%
1-Chloro-3-ethoxy-2-iodobenzene

Precaución: for Research Use Only

Formula: C₈H₈ClIO
MW: 282.5059 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34761446
CAS Number: 2807456-52-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR