2-Bromo-1-chloro-3-ethoxybenzene ≥97%

Supplier: AOBChem USA

Warning

63600-500MG 63600-1G 63600-5G 63600-25G
77331-942EA 221.45 USD
77331-942 77356-292 77368-044 77389-386
2-Bromo-1-chloro-3-ethoxybenzene ≥97%
2-Bromo-1-chloro-3-ethoxybenzene

Precaución: for Research Use Only

Formula: C₈H₈BrClO
Storage Temperature: Ambient
MDL Number: MFCD21609665
CAS Number: 1365271-74-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR