2,4-Dibromo-5-chlorophenol ≥97%

Supplier: AOBChem USA

Warning

63596-500MG 63596-1G 63596-5G
77335-878EA 367.71 USD
77335-878 77356-290 77380-298
2,4-Dibromo-5-chlorophenol ≥97%
2,4-Dibromo-5-chlorophenol

Precaución: for Research Use Only

Formula: C₆H₃Br₂ClO
Storage Temperature: Ambient
MDL Number: MFCD30661265
CAS Number: 89284-48-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR