5-Bromo-4-chloro-2-methoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

63587-500MG 63587-1G 63587-5G 63587-25G
77331-938EA 167.89 USD
77331-938 77346-224 77368-040 77389-384
5-Bromo-4-chloro-2-methoxybenzaldehyde ≥97%
5-Bromo-4-chloro-2-methoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₆BrClO₂
Storage Temperature: Ambient
MDL Number: MFCD28968004
CAS Number: 223778-51-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR