(2-Chloro-3-fluoro-4-isopropoxyphenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

63568-500MG 63568-1G 63568-5G
77331-936EA 342.99 USD
77331-936 77356-288 77380-294
(2-Chloro-3-fluoro-4-isopropoxyphenyl)(methyl)sulfane ≥97%
(2-Chloro-3-fluoro-4-isopropoxyphenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₁₀H₁₂ClFOS
Storage Temperature: Ambient
MDL Number: MFCD34761425
CAS Number: 2807469-86-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR