1-(Benzyloxy)-3-chloro-2-fluoro-4-iodobenzene ≥97%

Supplier: AOBChem USA

Warning

63556-500MG 63556-1G 63556-5G
77338-578EA 500.58 USD
77338-578 77356-284 77380-290
1-(Benzyloxy)-3-chloro-2-fluoro-4-iodobenzene ≥97%
1-(Benzyloxy)-3-chloro-2-fluoro-4-iodobenzene

Precaución: for Research Use Only

Formula: C₁₃H₉ClFIO
MW: 362.5658 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34761414
CAS Number: 2807450-43-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR