(4-(Benzyloxy)-2-chloro-3-fluorophenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

63555-500MG 63555-1G 63555-5G
77337-810EA 445.99 USD
77337-810 77356-282 77380-288
(4-(Benzyloxy)-2-chloro-3-fluorophenyl)(methyl)sulfane ≥97%
(4-(Benzyloxy)-2-chloro-3-fluorophenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₁₄H₁₂ClFOS
MW: 282.7608 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28503618
CAS Number: 1934380-02-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR