4-(Benzyloxy)-2-chloro-3-fluorobenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

63549-500MG 63549-1G 63549-5G
77339-272EA 571.65 USD
77339-272 77356-280 77380-286
4-(Benzyloxy)-2-chloro-3-fluorobenzaldehyde ≥97%
4-(Benzyloxy)-2-chloro-3-fluorobenzaldehyde

Precaución: for Research Use Only

Formula: C₁₄H₁₀ClFO₂
MW: 264.6794 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28503603
CAS Number: 1936692-17-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR