4-(Benzyloxy)-2-chloro-3-fluorobenzoic acid ≥97%

Supplier: AOBChem USA

Warning

63546-500MG 63546-1G 63546-5G
77337-204EA 408.91 USD
77337-204 77356-278 77380-284
4-(Benzyloxy)-2-chloro-3-fluorobenzoic acid ≥97%
4-(Benzyloxy)-2-chloro-3-fluorobenzoic acid

Precaución: for Research Use Only

Formula: C₁₄H₁₀ClFO₃
Storage Temperature: Ambient
MDL Number: MFCD28503598
CAS Number: 2384555-20-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR