1-(Benzyloxy)-4-bromo-3-chloro-2-fluorobenzene ≥95%

Supplier: AOBChem USA

Warning

63537-500MG 63537-1G 63537-5G 63537-25G
77331-934EA 198.79 USD
77331-934 77356-274 77368-038 77389-382
1-(Benzyloxy)-4-bromo-3-chloro-2-fluorobenzene ≥95%
1-(Benzyloxy)-4-bromo-3-chloro-2-fluorobenzene

Precaución: for Research Use Only

Formula: C₁₃H₉BrClFO
MW: 315.5653 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29044296
CAS Number: 1881330-85-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR