3-Bromo-2-ethoxy-5-isobutylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

63519-500MG 63519-1G
77340-488EA 1262.78 USD
77340-488 77356-266
3-Bromo-2-ethoxy-5-isobutylbenzaldehyde ≥97%
3-Bromo-2-ethoxy-5-isobutylbenzaldehyde

Precaución: for Research Use Only

Formula: C₁₃H₁₇BrO₂
MW: 285.1768 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34761397
CAS Number: 2807440-55-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR