2-(Benzyloxy)-5-isobutylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

63510-500MG 63510-1G 63510-5G
77339-086EA 548.99 USD
77339-086 77356-260 77380-272
2-(Benzyloxy)-5-isobutylbenzaldehyde ≥97%
2-(Benzyloxy)-5-isobutylbenzaldehyde

Precaución: for Research Use Only

Formula: C₁₈H₂₀O₂
Storage Temperature: Ambient
MDL Number: MFCD34761388
CAS Number: 2901066-02-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR