2-(2-Fluoro-5-methoxyphenyl)propan-2-ol ≥97%

Supplier: AOBChem USA

Warning

63476-500MG 63476-1G 63476-5G 63476-25G
77331-930EA 152.44 USD
77331-930 77345-484 77368-034 77389-378
2-(2-Fluoro-5-methoxyphenyl)propan-2-ol ≥97%
2-(2-Fluoro-5-methoxyphenyl)propan-2-ol

Precaución: for Research Use Only

Formula: C₁₀H₁₃FO₂
MW: 184.2074 g/mol
Storage Temperature: Ambient
MDL Number: MFCD27935125
CAS Number: 96826-29-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR