1-(3-(Benzyloxy)-2-bromo-6-fluorophenyl)-2,2,2-trifluoroethanone ≥97%

Supplier: AOBChem USA

Warning

63466-500MG 63466-1G 63466-5G 63466-25G
77331-924EA 258.53 USD
77331-924 77356-246 77368-028 77389-372
1-(3-(Benzyloxy)-2-bromo-6-fluorophenyl)-2,2,2-trifluoroethanone ≥97%
1-(3-(Benzyloxy)-2-bromo-6-fluorophenyl)-2,2,2-trifluoroethanone

Precaución: for Research Use Only

Formula: C₁₅H₉BrF₄O₂
MW: 377.1283 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34761359
CAS Number: 2807448-56-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR