1-(2,6-Difluoro-3-methoxyphenyl)-2,2,2-trifluoroethanone ≥95%

Supplier: AOBChem USA

Warning

63462-500MG 63462-1G 63462-5G 63462-25G
77331-922EA 263.68 USD
77331-922 77356-244 77368-026 77389-370
1-(2,6-Difluoro-3-methoxyphenyl)-2,2,2-trifluoroethanone ≥95%
1-(2,6-Difluoro-3-methoxyphenyl)-2,2,2-trifluoroethanone

Precaución: for Research Use Only

Formula: C₉H₅F₅O₂
MW: 240.1268 g/mol
Storage Temperature: Ambient
MDL Number: MFCD26687739
CAS Number: 1521210-39-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR