1-(3,6-Dichloro-2-fluorophenyl)-2,2,2-trifluoroethanone ≥97%
Supplier: AOBChem USA
|
Warning
|
63458-500MG
63458-1G
63458-5G
63458-25G
77331-918EA
257.5
USD
77331-918
77356-240
77368-022
77389-366
1-(3,6-Dichloro-2-fluorophenyl)-2,2,2-trifluoroethanone ≥97%
1-(3,6-Dichloro-2-fluorophenyl)-2,2,2-trifluoroethanone
Precaución: for Research Use Only
Formula:
C₈H₂Cl₂F₄O MW: 261.0004 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD34761358 CAS Number: 2807456-58-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...