1-(2,3-Dichloro-6-fluorophenyl)-2,2,2-trifluoroethanone ≥97%
Supplier: AOBChem USA
|
Warning
|
63457-500MG
63457-1G
63457-5G
63457-25G
77331-916EA
258.53
USD
77331-916
77356-238
77368-020
77389-364
1-(2,3-Dichloro-6-fluorophenyl)-2,2,2-trifluoroethanone ≥97%
1-(2,3-Dichloro-6-fluorophenyl)-2,2,2-trifluoroethanone
Precaución: for Research Use Only
Formula:
C₈H₂Cl₂F₄O MW: 261.0004 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD26687744 CAS Number: 1520481-67-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...