3-Fluoro-4-iodo-2-(methylthio)phenol ≥97%

Supplier: AOBChem USA

Warning

63450-500MG 63450-1G 63450-5G
77339-808EA 649.93 USD
77339-808 77356-232 77380-260
3-Fluoro-4-iodo-2-(methylthio)phenol ≥97%
3-Fluoro-4-iodo-2-(methylthio)phenol

Precaución: for Research Use Only

Formula: C₇H₆FIOS
MW: 284.0898 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34761357
CAS Number: 2707949-54-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR