2-Fluoro-4-hydroxy-3-(methylthio)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

63444-500MG 63444-1G 63444-5G
77338-308EA 473.8 USD
77338-308 77356-230 77380-258
2-Fluoro-4-hydroxy-3-(methylthio)benzoic acid ≥97%
2-Fluoro-4-hydroxy-3-(methylthio)benzoic acid

Precaución: for Research Use Only

Formula: C₈H₇FO₃S
MW: 202.2027 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34761351
CAS Number: 1891234-82-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR