1,5-Dibromo-2-iodo-3-(trifluoromethoxy)benzene ≥97%

Supplier: AOBChem USA

Warning

63437-500MG 63437-1G 63437-5G 63437-25G
77331-910EA 85.49 USD
77331-910 77342-230 77368-016 77380-254
1,5-Dibromo-2-iodo-3-(trifluoromethoxy)benzene ≥97%
1,5-Dibromo-2-iodo-3-(trifluoromethoxy)benzene

Precaución: for Research Use Only

Formula: C₇H₂Br₂F₃IO
Storage Temperature: Ambient
MDL Number: MFCD09878141
CAS Number: 1000578-23-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR