8-Chloro-5-fluoroquinoline ≥97%

Supplier: AOBChem USA

Warning

63430-500MG 63430-1G 63430-5G
77331-908EA 309 USD
77331-908 77356-226 77380-250
8-Chloro-5-fluoroquinoline ≥97%
8-Chloro-5-fluoroquinoline

Precaución: for Research Use Only

Formula: C₉H₅ClFN
MW: 181.5941 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09907836
CAS Number: 917251-76-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR