2-Chloro-1,3-difluoro-4-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

63428-500MG 63428-1G 63428-5G 63428-25G
77331-906EA 176.13 USD
77331-906 77346-560 77368-012 77389-360
2-Chloro-1,3-difluoro-4-methylbenzene ≥97%
2-Chloro-1,3-difluoro-4-methylbenzene

Precaución: for Research Use Only

Formula: C₇H₅ClF₂
MW: 162.5644 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12922568
CAS Number: 1208078-10-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR