3-Bromo-2-iodo-5-methylpyridine ≥97%

Supplier: AOBChem USA

Warning

63424-500MG 63424-1G 63424-5G 63424-25G
77331-904EA 184.37 USD
77331-904 77347-258 77368-010 77389-358
3-Bromo-2-iodo-5-methylpyridine ≥97%
3-Bromo-2-iodo-5-methylpyridine

Precaución: for Research Use Only

Formula: C₆H₅BrIN
Storage Temperature: Ambient
MDL Number: MFCD16610048
CAS Number: 1211542-16-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR