4-Ethoxy-1-iodo-2-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

63423-500MG 63423-1G 63423-5G 63423-25G
77331-902EA 120.51 USD
77331-902 77343-778 77368-008 77380-248
4-Ethoxy-1-iodo-2-methylbenzene ≥97%
4-Ethoxy-1-iodo-2-methylbenzene

Precaución: for Research Use Only

Formula: C₉H₁₁IO
MW: 262.0875 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32649094
CAS Number: 108178-36-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR