(4-Ethoxy-2-methylphenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

63422-500MG 63422-1G 63422-5G 63422-25G
77331-900EA 258.53 USD
77331-900 77356-224 77368-006 77389-356
(4-Ethoxy-2-methylphenyl)(methyl)sulfane ≥97%
(4-Ethoxy-2-methylphenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₁₀H₁₄OS
Storage Temperature: Ambient
MDL Number: MFCD22370316
CAS Number: 1783-80-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR