5-Bromo-2-(difluoromethoxy)-1-fluoro-3-methoxybenzene ≥97%

Supplier: AOBChem USA

Warning

63400-500MG 63400-1G 63400-5G
77340-066EA 741.6 USD
77340-066 77356-220 77380-244
5-Bromo-2-(difluoromethoxy)-1-fluoro-3-methoxybenzene ≥97%
5-Bromo-2-(difluoromethoxy)-1-fluoro-3-methoxybenzene

Precaución: for Research Use Only

Formula: C₈H₆BrF₃O₂
MW: 271.0312 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34761334
CAS Number: 2092778-93-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR