5-Bromo-3-fluoro-2-methoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

63379-500MG 63379-1G 63379-5G
77331-898EA 332.69 USD
77331-898 77356-216 77380-240
5-Bromo-3-fluoro-2-methoxybenzoic acid ≥97%
5-Bromo-3-fluoro-2-methoxybenzoic acid

Precaución: for Research Use Only

Formula: C₈H₆BrFO₃
Storage Temperature: Ambient
MDL Number: MFCD28063026
CAS Number: 1784034-92-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR