5-(3-Ethoxy-5-(trifluoromethyl)phenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

63375-500MG 63375-1G 63375-5G
77339-510EA 594.31 USD
77339-510 77356-212 77380-236
5-(3-Ethoxy-5-(trifluoromethyl)phenyl)thiazol-2-amine ≥97%
5-(3-Ethoxy-5-(trifluoromethyl)phenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₂H₁₁F₃N₂OS
MW: 288.2887 g/mol
Storage Temperature: Ambient
CAS Number: 2807469-87-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR