5-(3-Methyl-5-(methylthio)phenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

63364-500MG 63364-1G 63364-25G 63364-5G
77331-896EA 301.79 USD
77331-896 77356-208 77389-354 77380-232
5-(3-Methyl-5-(methylthio)phenyl)thiazol-2-amine ≥97%
5-(3-Methyl-5-(methylthio)phenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₁H₁₂N₂S₂
MW: 236.3563 g/mol
Storage Temperature: Ambient
CAS Number: 2807442-99-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR