2,2,2-trifluoro-1-(3-hydroxyphenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

63353-500MG 63353-1G 63353-5G
77340-164EA 806.49 USD
77340-164 77356-206 77380-230
2,2,2-trifluoro-1-(3-hydroxyphenyl)ethanone ≥95%
2,2,2-trifluoro-1-(3-hydroxyphenyl)ethanone

Precaución: for Research Use Only

Formula: C₈H₅F₃O₂
MW: 190.1193 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13172697
CAS Number: 326897-98-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR