2-Fluoro-3-(methylthio)terephthalaldehyde ≥97%

Supplier: AOBChem USA

Warning

63319-500MG 63319-1G 63319-5G
77338-024EA 459.38 USD
77338-024 77356-204 77380-228
2-Fluoro-3-(methylthio)terephthalaldehyde ≥97%
2-Fluoro-3-(methylthio)terephthalaldehyde

Precaución: for Research Use Only

Formula: C₉H₇FO₂S
MW: 198.2140 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34761290
CAS Number: 2090354-16-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR