3-Fluoro-4-iodo-2-(methylthio)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

63317-500MG 63317-1G 63317-5G
77338-906EA 528.39 USD
77338-906 77356-202 77380-226
3-Fluoro-4-iodo-2-(methylthio)benzaldehyde ≥97%
3-Fluoro-4-iodo-2-(methylthio)benzaldehyde

Precaución: for Research Use Only

Formula: C₈H₆FIOS
MW: 296.1005 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34761288
CAS Number: 2091006-32-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR