5-Bromo-2-ethoxy-3-fluorobenzoic acid ≥97%

Supplier: AOBChem USA

Warning

63307-500MG 63307-1G 63307-5G
77336-528EA 388.31 USD
77336-528 77356-194 77380-218
5-Bromo-2-ethoxy-3-fluorobenzoic acid ≥97%
5-Bromo-2-ethoxy-3-fluorobenzoic acid

Precaución: for Research Use Only

Formula: C₉H₈BrFO₃
Storage Temperature: Ambient
MDL Number: MFCD28063048
CAS Number: 1784193-74-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR