1-(2-Bromo-6-fluoro-4-methylphenyl)-2,2,2-trifluoroethanone ≥97%

Supplier: AOBChem USA

Warning

63291-500MG 63291-1G 63291-5G
77340-158EA 800.31 USD
77340-158 77356-186 77380-212
1-(2-Bromo-6-fluoro-4-methylphenyl)-2,2,2-trifluoroethanone ≥97%
1-(2-Bromo-6-fluoro-4-methylphenyl)-2,2,2-trifluoroethanone

Precaución: for Research Use Only

Formula: C₉H₅BrF₄O
MW: 285.0330 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34761276
CAS Number: 2807446-48-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR