1-(6-Bromo-3-chloro-2-fluorophenyl)-2,2,2-trifluoroethanone ≥97%
Supplier: AOBChem USA
|
Warning
|
63288-500MG
63288-1G
63288-5G
77340-092EA
753.96
USD
77340-092
77356-184
77380-210
1-(6-Bromo-3-chloro-2-fluorophenyl)-2,2,2-trifluoroethanone ≥97%
1-(6-Bromo-3-chloro-2-fluorophenyl)-2,2,2-trifluoroethanone
Precaución: for Research Use Only
Formula:
C₈H₂BrClF₄O MW: 305.4514 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD28141986 CAS Number: 1823493-67-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...