1-(3-Bromo-2,6-difluorophenyl)-2,2,2-trifluoroethanone ≥95%

Supplier: AOBChem USA

Warning

63287-500MG 63287-1G 63287-5G 63287-25G
77331-886EA 263.68 USD
77331-886 77356-182 77367-996 77389-344
1-(3-Bromo-2,6-difluorophenyl)-2,2,2-trifluoroethanone ≥95%
1-(3-Bromo-2,6-difluorophenyl)-2,2,2-trifluoroethanone

Precaución: for Research Use Only

Formula: C₈H₂BrF₅O
Storage Temperature: Ambient
MDL Number: MFCD12922601
CAS Number: 1208074-69-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR