1-(2-Bromo-3-chloro-6-fluorophenyl)-2,2,2-trifluoroethanone ≥97%

Supplier: AOBChem USA

Warning

63286-500MG 63286-1G 63286-5G
77340-090EA 753.96 USD
77340-090 77356-180 77380-208
1-(2-Bromo-3-chloro-6-fluorophenyl)-2,2,2-trifluoroethanone ≥97%
1-(2-Bromo-3-chloro-6-fluorophenyl)-2,2,2-trifluoroethanone

Precaución: for Research Use Only

Formula: C₈H₂BrClF₄O
MW: 305.4514 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28141961
CAS Number: 1823559-69-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR