1-(3,6-Dibromo-2-fluorophenyl)-2,2,2-trifluoroethanone ≥97%
Supplier: AOBChem USA
|
Warning
|
63277-500MG
63277-1G
63277-5G
63277-25G
77331-878EA
145.23
USD
77331-878
77345-106
77367-990
77389-336
1-(3,6-Dibromo-2-fluorophenyl)-2,2,2-trifluoroethanone ≥97%
1-(3,6-Dibromo-2-fluorophenyl)-2,2,2-trifluoroethanone
Precaución: for Research Use Only
Formula:
C₈H₂Br₂F₄O Storage Temperature: Ambient |
MDL Number:
MFCD34761271 CAS Number: 2764731-45-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...