5-Bromo-2-ethoxy-1-fluoro-3-iodobenzene ≥97%

Supplier: AOBChem USA

Warning

63271-500MG 63271-1G 63271-5G 63271-25G
77331-874EA 204.97 USD
77331-874 77356-172 77367-986 77389-332
5-Bromo-2-ethoxy-1-fluoro-3-iodobenzene ≥97%
5-Bromo-2-ethoxy-1-fluoro-3-iodobenzene

Precaución: for Research Use Only

Formula: C₈H₇BrFIO
MW: 344.9474 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34761268
CAS Number: 2386386-61-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR