1-(3,5-Dichloro-2-methylphenyl)-2,2,2-trifluoroethanol ≥97%

Supplier: AOBChem USA

Warning

63270-500MG 63270-1G 63270-5G
77331-872EA 330.63 USD
77331-872 77356-170 77380-202
1-(3,5-Dichloro-2-methylphenyl)-2,2,2-trifluoroethanol ≥97%
1-(3,5-Dichloro-2-methylphenyl)-2,2,2-trifluoroethanol

Precaución: for Research Use Only

Formula: C₉H₇Cl₂F₃O
MW: 259.0524 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34761267
CAS Number: 2901098-69-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR